accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSBr

CAS RN 934570-51-1

IUPAC Name: 4-(3-bromothiophen-2-yl)benzaldehyde
Synonyms: 4-(3-bromothiophen-2-yl)benzaldehyde
Molecular Weight (g/mol): 267.14
Molecular Formula: C11H7BrOS
InChi Key: BTYZFUJFFBNJTP-UHFFFAOYSA-N
SMILES: BrC1=C(SC=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results