accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHN

CAS RN 934570-59-9

IUPAC Name: {1-[(pyridin-3-yl)methyl]piperidin-4-yl}methanol
Synonyms: [1-(pyridin-3-ylmethyl)piperidin-4-yl]methanol
Molecular Weight (g/mol): 206.29
Molecular Formula: C12H18N2O
InChi Key: RAJQONMXWKPTTO-UHFFFAOYSA-N
SMILES: OCC1CCN(CC2=CN=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results