accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNNH2

CAS RN 935-69-3

IUPAC Name: 7-methyl-7H-purin-6-amine
Synonyms: 7-methyladenine
Molecular Weight (g/mol): 149.16
Molecular Formula: C6H7N5
InChi Key: HCGHYQLFMPXSDU-UHFFFAOYSA-N
SMILES: CN1C=NC2=NC=NC(N)=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)