accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClClCl

CAS RN 935-95-5

IUPAC Name: 2,3,5,6-tetrachlorophenol
Synonyms: 2,3,5,6-tetrachlorophenol 2,3,5,6-tetrachlorophenate C6H2cl4O
Molecular Weight (g/mol): 231.88
Molecular Formula: C6H2Cl4O
InChi Key: KEWNKZNZRIAIAK-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C(Cl)=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results