accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3POOOOPCH3CH3CH3H3CH3CCH3CH3CH3POOOOPCH3CH3CH3H3CH3CCH3CH3H3CNHCH3HClClClHRuClHHClRu(?)(?)

CAS RN 935449-46-0

IUPAC Name: bis((4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane) [chloraniumyl(chloro)ruthenio]chloranium dimethylamine hydrochloride ruthenium chloride
Synonyms: bis((4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane) [chloraniumyl(chloro)ruthenio]chloranium dimethylamine hydrochloride ruthenium chloride
Molecular Weight (g/mol): 1874.11
Molecular Formula: C94H99Cl5NO8P4Ru2
InChi Key: CABGTUSYWYURQC-UHFFFAOYSA-M
SMILES: Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results