accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 935521-01-0

IUPAC Name: N4-(2-methylpropyl)quinoline-3,4-diamine hydrochloride
Synonyms: N4-(2-methylpropyl)quinoline-3,4-diamine hydrochloride
Molecular Weight (g/mol): 251.76
Molecular Formula: C13H18ClN3
InChi Key: ZRRAINCPRZTHMN-UHFFFAOYSA-N
SMILES: Cl.CC(C)CNC1=C2C=CC=CC2=NC=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results