accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 936-49-2

IUPAC Name: 2-phenyl-4,5-dihydro-1H-imidazole
Synonyms: 2-phenyl-2-imidazoline
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: BKCCAYLNRIRKDJ-UHFFFAOYSA-N
SMILES: C1CN=C(N1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)