accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(S)(S)OH3CH2N

CAS RN 93621-94-4

IUPAC Name: (2S,5S)-2,5-bis(methoxymethyl)pyrrolidin-1-ium
Synonyms: (2S,5S)-2,5-bis(methoxymethyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 160.24
Molecular Formula: C8H18NO2
InChi Key: HDXYTIYVVNJFLU-YUMQZZPRSA-O
SMILES: COC[C@@H]1CC[C@@H](COC)[NH2+]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results