accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNNNH2HOOH

CAS RN 936250-22-5

IUPAC Name: (2-aminopyrimidin-5-yl)boronic acid
Synonyms: 2-aminopyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 138.92
Molecular Formula: C4H6BN3O2
InChi Key: CGHYQZASLKERLV-UHFFFAOYSA-N
SMILES: NC1=NC=C(C=N1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)