accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 936707-21-0

IUPAC Name: (5R)-3-[(2S)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (5R)-3-[(2S)-3-[1-(tert-butoxycarbonyl)indol-3-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 667.76
Molecular Formula: C38H41N3O8
InChi Key: XCLJOQSZEAUSDT-IDTHGENUSA-N
SMILES: C[C@H]1OC(C)(C)N(C1C(O)=O)C(=O)[C@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)