accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 937-20-2

IUPAC Name: 2-chloro-1-(4-chlorophenyl)ethan-1-one
Synonyms: 2,4'-dichloroacetophenone
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: FWDFNLVLIXAOMX-UHFFFAOYSA-N
SMILES: ClCC(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)