accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 937-63-3

IUPAC Name: 4-methylphenyl chloromethanethioate
Synonyms: 4-methylphenyl chloromethanethioate
Molecular Weight (g/mol): 186.65
Molecular Formula: C8H7ClOS
InChi Key: UNCAXIZUVRKBMN-UHFFFAOYSA-N
SMILES: CC1=CC=C(OC(Cl)=S)C=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)