accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOOHNH2N

CAS RN 937039-45-7

IUPAC Name: N'-(2-aminophenyl)-N-(4-methylphenyl)heptanediamide
Synonyms: N'-(2-aminophenyl)-N-(4-methylphenyl)heptanediamide
Molecular Weight (g/mol): 339.44
Molecular Formula: C20H25N3O2
InChi Key: WTKBRPXPNAKVEQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(=O)CCCCCC(=O)NC2=CC=CC=C2N)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results