CAS RN 937082-81-0
CAS RN 937082-81-0
IUPAC Name:
9-(4-bromophenyl)-9-phenyl-9H-fluorene
Synonyms:
9-(4-bromophenyl)-9-phenylfluorene
Molecular Weight (g/mol):
397.32
Molecular Formula:
C25H17Br
InChi Key:
OOKRYIPMHLUQHU-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
1
–
1
of
1
results
1
9-(4-Bromophenyl)-9-phenylfluorene 98.0+%, TCI America™
CAS: 937082-81-0 Molecular Formula: C25H17Br Molecular Weight (g/mol): 397.315 MDL Number: MFCD28975087 InChI Key: OOKRYIPMHLUQHU-UHFFFAOYSA-N PubChem CID: 57746345 IUPAC Name: 9-(4-bromophenyl)-9-phenylfluorene SMILES: C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)Br