accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 937796-01-5

IUPAC Name: 1-[4-(3-bromothiophen-2-yl)phenyl]ethan-1-one
Synonyms: 1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 281.17
Molecular Formula: C12H9BrOS
InChi Key: AMADTVLVNSCDKW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results