accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 937796-10-6

IUPAC Name: 2-(pyrrolidin-1-yl)pyrimidine-5-carbaldehyde
Synonyms: 2-(pyrrolidin-1-yl)pyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 177.21
Molecular Formula: C9H11N3O
InChi Key: KJMKROLYLRRJDN-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(N=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results