accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNHO

CAS RN 937796-13-9

IUPAC Name: [2-(morpholin-4-yl)pyrimidin-5-yl]methanol
Synonyms: [2-(morpholin-4-yl)pyrimidin-5-yl]methanol
Molecular Weight (g/mol): 195.22
Molecular Formula: C9H13N3O2
InChi Key: KYYQOJQTQOKGPQ-UHFFFAOYSA-N
SMILES: OCC1=CN=C(N=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results