accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 937796-15-1

IUPAC Name: [2-(morpholine-4-sulfonyl)phenyl]methanol
Synonyms: [2-(morpholine-4-sulfonyl)phenyl]methanol
Molecular Weight (g/mol): 257.3
Molecular Formula: C11H15NO4S
InChi Key: BOQHDNNWBAICEW-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results