accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNN

CAS RN 937796-17-3

IUPAC Name: methyl({1-[(pyridin-3-yl)methyl]piperidin-4-yl}methyl)amine
Synonyms: methyl({[1-(pyridin-3-ylmethyl)piperidin-4-yl]methyl})amine
Molecular Weight (g/mol): 219.33
Molecular Formula: C13H21N3
InChi Key: NGMNACILLKNUTM-UHFFFAOYSA-N
SMILES: CNCC1CCN(CC2=CN=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results