accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClCl

CAS RN 938-71-6

IUPAC Name: 1-chloro-3-(chloromethyl)-5-nitrobenzene
Synonyms: 1-chloro-3-(chloromethyl)-5-nitrobenzene
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: QBIQAXWMONJLNW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results