accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)OHNClBr(R)OH(S)HO(R)HO

CAS RN 93863-88-8

IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R,6R)-2-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 408.63
Molecular Formula: C14H15BrClNO6
InChi Key: CHRVKCMQIZYLNM-MBJXGIAVSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)