accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNNH2NSClOHOPOOONa

CAS RN 93882-12-3

IUPAC Name: sodium 6-{6-amino-8-[(4-chlorophenyl)sulfanyl]-9H-purin-9-yl}-7-hydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonyms: sodium 6-{6-amino-8-[(4-chlorophenyl)sulfanyl]purin-9-yl}-7-hydroxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Weight (g/mol): 493.79
Molecular Formula: C16H14ClN5NaO6PS
InChi Key: YIJFVHMIFGLKQL-UHFFFAOYNA-M
SMILES: [Na+].NC1=C2N=C(SC3=CC=C(Cl)C=C3)N(C3OC4COP([O-])(=O)OC4C3O)C2=NC=N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results