accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 939-23-1

IUPAC Name: 4-phenylpyridine
Synonyms: 4-phenylpyridine azabiphenyl
Molecular Weight (g/mol): 155.2
Molecular Formula: C11H9N
InChi Key: JVZRCNQLWOELDU-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=NC=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)