accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 939-69-5

IUPAC Name: 6-hydroxy-1,3-benzothiazole-2-carbonitrile
Synonyms: 6-hydroxy-1,3-benzothiazole-2-carbonitrile
Molecular Weight (g/mol): 176.19
Molecular Formula: C8H4N2OS
InChi Key: SQAVNBZDECKYOT-UHFFFAOYSA-N
SMILES: OC1=CC=C2N=C(SC2=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)