accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO(R)(R)

CAS RN 939-90-2

IUPAC Name: (1R,2R)-2-phenylcyclopropane-1-carboxylic acid
Synonyms: 2-PhCPCOOH
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: AHDDRJBFJBDEPW-DTWKUNHWSA-N
SMILES: OC(=O)[C@@H]1C[C@H]1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results