accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 939-99-1

FFFCl

CAS RN 939-99-1

IUPAC Name: 1-(chloromethyl)-4-(trifluoromethyl)benzene
Synonyms: 1-(chloromethyl)-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.58
Molecular Formula: C8H6ClF3
InChi Key: MCHDHQVROPEJJT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results