accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H

CAS RN 93951-69-0

IUPAC Name: (²H₁₀)fluoranthene
Synonyms: (²H₁₀)fluoranthene
Molecular Weight (g/mol): 212.32
Molecular Formula: C16H10
InChi Key: GVEPBJHOBDJJJI-LHNTUAQVSA-N
SMILES: [2H]C1=C([2H])C([2H])=C2C(=C1[2H])C1=C([2H])C([2H])=C([2H])C3=C([2H])C([2H])=C([2H])C2=C13

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results