accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 93951-97-4

IUPAC Name: acenaphthylene
Synonyms: acenaphthylene cyclopenta(de)naphthalene acenaphthalene
Molecular Weight (g/mol): 152.2
Molecular Formula: C12H8
InChi Key: HXGDTGSAIMULJN-UHFFFAOYSA-N
SMILES: C1=CC2=C3C1=CC=CC3=CC=C2
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results