CAS RN 93952-14-8
IUPAC Name:
hexachlorobenzene
Synonyms:
hexachlorobenzene HCB phenyl perchloryl
Molecular Weight (g/mol):
284.77
Molecular Formula:
C6Cl6
InChi Key:
CKAPSXZOOQJIBF-UHFFFAOYSA-N
SMILES:
ClC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl