accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94-04-2

IUPAC Name: ethenyl 2-ethylhexanoate
Synonyms: ethenyl 2-ethylhexanoate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: IGBZOHMCHDADGY-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(=O)OC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results