accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH2

CAS RN 94-09-7

IUPAC Name: ethyl 4-aminobenzoate
Synonyms: benzocaine|americaine|hurricaine|chloraseptic|dermoplast|ethyl P-aminobenzoate|ethyl aminobenzoate|ora-jel|Ora-Jel|baby anbesol
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(N)C=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (6)
  • (6)
Physical Form 
  • (1)
  • (5)
  • (6)
Boiling Point 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (1)
  • (6)
  • (1)
  • (2)
  • (6)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (6)
  • (1)
  • (5)
  • (6)
  • (3)
  • (5)
  • (6)
  • (1)