accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94-34-8

IUPAC Name: 3-[methyl(phenyl)amino]propanenitrile
Synonyms: 3-[methyl(phenyl)amino]propanenitrile
Molecular Weight (g/mol): 160.22
Molecular Formula: C10H12N2
InChi Key: IXXLKTZOCSRXEM-UHFFFAOYSA-N
SMILES: CN(CCC#N)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results