accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOO

CAS RN 94-49-5

IUPAC Name: 2-(benzoyloxy)ethyl benzoate
Synonyms: 1,2-ethanediol, dibenzoate
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: XFDQLDNQZFOAFK-UHFFFAOYSA-N
SMILES: O=C(OCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results