accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNO

CAS RN 94-69-9

IUPAC Name: N-(2-methylphenyl)formamide
Synonyms: formamide, N-(2-methylphenyl)-
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: ZXTLGJAARBNQGK-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results