accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH2N

CAS RN 94-70-2

IUPAC Name: 2-ethoxyaniline
Synonyms: 2-ethoxyaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ULHFFAFDSSHFDA-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1N
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)