accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOClCl

CAS RN 94-75-7

IUPAC Name: 2-(2,4-dichlorophenoxy)acetic acid
Synonyms: rhodia pennamine D miracle netagrone emulsamine E-3 brush-rhap scott's 4-XD weed control caswell no. 315 green cross weed-no-more "80" ipaner
Molecular Weight (g/mol): 221.03
Molecular Formula: C8H6Cl2O3
InChi Key: OVSKIKFHRZPJSS-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (9)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (9)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)