accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOClCl

CAS RN 94-82-6

IUPAC Name: 4-(2,4-dichlorophenoxy)butanoic acid
Synonyms: butyrac sys 67 buratal caswell no. 316 butyrac 200 butyrac 118 butoxon 2,4-D butyric acid butyrac ester butoxone ester butirex
Molecular Weight (g/mol): 249.09
Molecular Formula: C10H10Cl2O3
InChi Key: YIVXMZJTEQBPQO-UHFFFAOYSA-N
SMILES: OC(=O)CCCOC1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results