accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3HO(E)

CAS RN 94-86-0

IUPAC Name: 2-ethoxy-5-[(1E)-prop-1-en-1-yl]phenol
Synonyms: ethoxy-propenylphenol
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: RADIRXJQODWKGQ-HWKANZROSA-N
SMILES: CCOC1=CC=C(\C=C\C)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results