accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHO

CAS RN 940-64-7

IUPAC Name: 2-(4-methylphenoxy)acetic acid
Synonyms: P-methylphenoxyacetic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: SFTDDFBJWUWKMN-UHFFFAOYSA-N
SMILES: CC1=CC=C(OCC(O)=O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)