accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94035-02-6

IUPAC Name: 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol; tetrakis(propan-1-ol)
Synonyms: cyclo-hepta-amylose; tetrakis(propanol)
Molecular Weight (g/mol): 1375.37
Molecular Formula: C54H102O39
InChi Key: BZRWCHBEBFPRGF-UHFFFAOYNA-N
SMILES: CCCO.CCCO.CCCO.CCCO.OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results