accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3Br

CAS RN 941-37-7

IUPAC Name: 1-bromo-3,5-dimethyladamantane
Synonyms: 1-bromo-3,5-dimethyladamantane
Molecular Weight (g/mol): 243.19
Molecular Formula: C12H19Br
InChi Key: QUCXLVDIVQWYJR-UHFFFAOYNA-N
SMILES: CC12CC3CC(C)(C1)CC(Br)(C3)C2
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)