accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94124-39-7

IUPAC Name: bromo(tert-butyl)methoxy(phenyl)silane
Synonyms: bromo(tert-butyl)methoxy(phenyl)silane
Molecular Weight (g/mol): 273.25
Molecular Formula: C11H17BrOSi
InChi Key: VFFLZSQFWJXUJJ-UHFFFAOYNA-N
SMILES: CO[Si](Br)(C1=CC=CC=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results