accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNCO

CAS RN 941716-86-5

IUPAC Name: 5-(3-isocyanatophenyl)-1-methyl-1H-pyrazole
Synonyms: 5-(3-isocyanatophenyl)-1-methylpyrazole
Molecular Weight (g/mol): 199.21
Molecular Formula: C11H9N3O
InChi Key: ZFRFBJVFEQIMLM-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C1=CC(=CC=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results