accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNSOOH

CAS RN 941716-91-2

IUPAC Name: 2-(pyrrolidin-1-yl)-1,3-thiazole-5-carboxylic acid
Synonyms: 2-(pyrrolidin-1-yl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 198.24
Molecular Formula: C8H10N2O2S
InChi Key: KPQGRYOIUITVHX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(S1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results