accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NHNCl

CAS RN 942-26-7

IUPAC Name: hydrogen 2-(5-chloro-1H-indol-3-yl)ethan-1-amine chloride
Synonyms: hydrogen 2-(5-chloro-1H-indol-3-yl)ethanamine chloride
Molecular Weight (g/mol): 231.12
Molecular Formula: C10H12Cl2N2
InChi Key: PBANXRNIXGEHPZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CNC2=CC=C(Cl)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results