accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOONOOOGd

CAS RN 94219-55-3

IUPAC Name: gadolinium(3+) trinitrate
Synonyms: gadolinium(III) nitrate
Molecular Weight (g/mol): 343.26
Molecular Formula: GdN3O9
InChi Key: MWFSXYMZCVAQCC-UHFFFAOYSA-N
SMILES: [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)