CAS RN 943-45-3
IUPAC Name:
2-methyl-2-phenoxypropanoic acid
Synonyms:
phenoxyisobutyric acid
Molecular Weight (g/mol):
180.2
Molecular Formula:
C10H12O3
InChi Key:
ILPUOPPYSQEBNJ-UHFFFAOYSA-N
SMILES:
CC(C)(OC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
2-Methyl-2-phenoxypropionic Acid 98.0+%, TCI America™
CAS: 943-45-3 Molecular Formula: C10H12O3 Molecular Weight (g/mol): 180.203 MDL Number: MFCD00129939 InChI Key: ILPUOPPYSQEBNJ-UHFFFAOYSA-N Synonym: 2-Phenoxyisobutyric Acid PubChem CID: 70343 IUPAC Name: 2-methyl-2-phenoxypropanoic acid SMILES: CC(C)(C(=O)O)OC1=CC=CC=C1