accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94347-11-2

IUPAC Name: 4-benzyl 1-tert-butyl 2-aminobutanedioate hydrochloride
Synonyms: 4-benzyl 1-tert-butyl 2-aminobutanedioate hydrochloride
Molecular Weight (g/mol): 315.79
Molecular Formula: C15H22ClNO4
InChi Key: YSDLFDMXCRFHTQ-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)C(N)CC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)