accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 943911-64-6

IUPAC Name: N-[2-(morpholin-4-yl)ethyl]-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: N-[2-(morpholin-4-yl)ethyl]-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 333.24
Molecular Formula: C17H28BN3O3
InChi Key: ZYQJAKOVUNNVAU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(NCCN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results