accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944-22-9

H3COPSCH3S

CAS RN 944-22-9

IUPAC Name: ethoxy(ethyl)(phenylsulfanyl)-λ⁵-phosphanethione
Synonyms: doubledown difonatul stauffer n-2790 fonofos (substance) stauffer N2790 caswell no. 454B difonate fonophos stauffer N-2790 stauffer n 2790
Molecular Weight (g/mol): 246.32
Molecular Formula: C10H15OPS2
InChi Key: KVGLBTYUCJYMND-UHFFFAOYNA-N
SMILES: CCOP(=S)(CC)SC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results